Learn More
Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01310509
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel(II) telluride | |
Lump | |
(metals basis excluding Co), Co 0.1 to 1% | |
NiTe | |
UN3077 | |
NPEUSMKUOOTUGX-UHFFFAOYSA-N | |
tellanylidenenickel | |
62780 | |
99.9% |
12142-88-0 | |
10 g | |
NiTe | |
MFCD00016258 | |
nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickeltellurid | |
[Ni]=[Te] | |
186.29 | |
186.31 | |
Odorless |
Chemical Identifiers
12142-88-0 | |
186.29 | |
NPEUSMKUOOTUGX-UHFFFAOYSA-N | |
62780 | |
[Ni]=[Te] |
NiTe | |
MFCD00016258 | |
nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickeltellurid | |
tellanylidenenickel |
Safety and Handling
GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350i-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 235-260-6
RTECSNumber : QS0400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only