missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%

Manufacturer:  Alfa Aesar™ 1310518

 View more versions of this product

Catalog No. AA1310518



Specifications

Specifications

Nickel(II) telluride
(metals basis excluding Co), Co 0.1 to 1%
NiTe
UN3077
NPEUSMKUOOTUGX-UHFFFAOYSA-N
tellanylidenenickel
62780
Lumps
Odorless
50g
12142-88-0
NiTe
MFCD00016258
nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickel telluride, nickeltellurid
[Ni]=[Te]
186.293
186.31
99.9%
10mm & down

Chemical Identifiers

12142-88-0
186.293
NPEUSMKUOOTUGX-UHFFFAOYSA-N
62780
[Ni]=[Te]
NiTe
MFCD00016258
nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickel telluride, nickeltellurid
tellanylidenenickel
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : QS0400000

TSCA : TSCA

Documents