missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis)

Manufacturer:  Alfa Aesar™ 1082022

 View more versions of this product

Catalog No. AA1082022



Specifications

Specifications

Nickel(II) sulfate hydrate
(metals basis)
NiSO4·xH2O (x≈6)
UN3288
nickel 2+ sulfate
[O-]S(=O)(=O)[O-].[Ni+2]
154.749
CHEBI:53001
Crystalline Powder
Puratronic™
Odorless
4.3 to 4.7
15244-37-8
NiO4S
MFCD00011146
14,6517
LGQLOGILCSXPEA-UHFFFAOYSA-L
nickel(2+);sulfate
24586
154.77 (Anhydrous)
99.9985%
2.070g/mL
Green
100g

Chemical Identifiers

15244-37-8
154.749
LGQLOGILCSXPEA-UHFFFAOYSA-L
24586
nickel(2+);sulfate
NiO4S
MFCD00011146
nickel 2+ sulfate
CHEBI:53001
[O-]S(=O)(=O)[O-].[Ni+2]
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P273-P201-P309+P311-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents