Learn More
Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 211080100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nickel(II) sulfate hexahydrate | |
10101-97-0,7786-81-4 | |
100.0 | |
98.5% min. (Complexometry) | |
NiSO4·6H2O | |
15,6602 | |
Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
262.85 | |
CHEBI:53437 | |
99% | |
Plastic drum |
99% | |
98.5 | |
10kg | |
NiO4S·6H2O | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
nickel(2+);sulfate;hexahydrate | |
5284429 | |
262.85 | |
Analysis |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
M
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
GHS Signal Word: Danger
RTECSNumber : QR9600000
RUO â Research Use Only