missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC270552500

 View more versions of this product

Catalog No. AC270552500



Specifications

Specifications

Nickel(II) sulfate heptahydrate
NiO47H2O
MFCD00149814
ksc492q3n
Solubility in water: 625g/L (20°C). Other solubilities: insoluble in alcohol and ether
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
280.88
CHEBI:53504
Needle-Like Crystals or Powder
Analysis
5000ppm max.
0.01% max. (in 0.1 N H2SO4)
10ppm max.
30ppm max.
250g
10101-98-1,7786-81-4
NiSO4·7H2O
15, 6602
10ppm max.
OGKAGKFVPCOHQW-UHFFFAOYSA-L
nickel(2+);sulfate;heptahydrate
61474
280.88
20 to 21% (Ni)
10ppm max.
20ppm max.
20ppm max.
Plastic bottle
Green

Chemical Identifiers

10101-98-1
280.88
OGKAGKFVPCOHQW-UHFFFAOYSA-L
61474
nickel(2+);sulfate;heptahydrate
NiO47H2O
MFCD00149814
ksc492q3n
CHEBI:53504
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
M

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

GHS Signal Word: Danger

SDS Documents