Please login to your online account to display your discounted pricing

Nickel(II) sulfamate hydrate, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Nickel(II) sulfamate hydrate
CAS 13770-89-3
Molecular Formula Ni(SO3NH2)2·xH2O
Formula Weight 250.86 (Anhydrous)
MDL Number MFCD00137261
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA8902030

  SDS
alfa aesar
8902030
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.86 (Anhydrous)
Quantity 250g
Molecular Formula Ni(SO3NH2)2·xH2O
Health Hazard 1 Danger
CAS 13770-89-3
RTECS Number QR9275000
Health Hazard 3 GHS P Statement
P260-P285-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Color Blue
Physical Form Crystalline
MDL Number MFCD00137261
Chemical Name or Material Nickel(II) sulfamate hydrate
Health Hazard 2 GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA TSCA
UN Number UN3077
Safety and Handling

Danger

GHS P Statement
P260-P285-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.