missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) sulfamate hydrate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 08902030

 View more versions of this product

Catalog No. AA8902030



Description

Description

Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel(II) sulfamate hydrate
Blue
250 g
MFCD00137261
nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
KERTUBUCQCSNJU-UHFFFAOYSA-L
nickel(2+);disulfamate
83720
Odorless
13770-89-3
Crystalline
H4N2NiO6S2
UN3077
Soluble in water.
[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O
250.85
250.86 (Anhydrous)

Chemical Identifiers

13770-89-3
250.85
KERTUBUCQCSNJU-UHFFFAOYSA-L
83720
H4N2NiO6S2
MFCD00137261
nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H341-H350i-H360D-H372

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 237-396-1

RTECSNumber : QR9275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only