Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) stearate

Chemical Identifiers

Chemical Name or Material Nickel(II) stearate
CAS 2223-95-2
Formula Weight 625.67g/mol
MDL Number MFCD00054344
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3945122

 
alfa aesar™
39451-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3945136

 
alfa aesar™
39451-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39451A3

 
alfa aesar™
39451-A3
2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100g
Density 1.13g/mL
Quantity 100g
Odor Odorless
Chemical Name or Material Nickel(II) stearate
CAS 2223-95-2
Physical Form Powder
Formula Weight 625.67g/mol
MDL Number MFCD00054344
UN Number UN3077
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeat
Health Hazard 3 GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store l
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store l

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeat