Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) selenide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Nickel(II) selenide
CAS 1314-05-2
Formula Weight 137.67
MDL Number MFCD00049497
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1310606

  SDS
alfa aesar™
1310606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1310614

  SDS
alfa aesar™
1310614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 137.67
Chemical Name or Material Nickel(II) selenide
Odor Odorless
Particle Size 6mm & down
Physical Form Pieces
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00049497
CAS 1314-05-2
UN Number UN3077
Quantity 5g
Solubility Information Insoluble in water; Soluble in aqua regia, HNO3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 8.46g/mL
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number QR8926000
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.