Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) phosphate hydrate

Chemical Identifiers

Chemical Name or Material Nickel(II) phosphate hydrate
CAS 27176-17-6
Formula Weight 366.01 (Anhydrous)
MDL Number MFCD00049501
UN Number UN3077
Merck Index 14,6515
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39478A1

 
alfa aesar™
39478A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 366.01 (Anhydrous)
Chemical Name or Material Nickel(II) phosphate hydrate
TSCA TSCA
Physical Form Lump/Powder
MDL Number MFCD00049501
CAS 27176-17-6
UN Number UN3077
Merck Index 14,6515
Color Green
Quantity 1kg
Solubility Information Insoluble in water; Soluble in acids, ammonia
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H350-H372-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store l
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store l

GHS H Statement
H334-H350-H372-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.