missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) perchlorate hydrate, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 4431609

 View more versions of this product

Catalog No. AA4431609



Specifications

Specifications

Nickel (II) perchlorate hydrate
(metals basis)
Ni(ClO4)2·xH2O
UN1481
Very soluble in water. Soluble in many organic solvents.
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2]
257.585
257.59 (Anhydrous)
99.998%
1.55
Green
10g
13637-71-3
Cl2NiO8
MFCD00149804
bq8a24mezo
ZLQBNKOPBDZKDP-UHFFFAOYSA-L
nickel(2+);diperchlorate
26158
Crystalline
Odorless
Hygroscopic
140°C

Chemical Identifiers

13637-71-3
257.585
ZLQBNKOPBDZKDP-UHFFFAOYSA-L
26158
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2]
Cl2NiO8
MFCD00149804
bq8a24mezo
nickel(2+);diperchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H334-H350-H360-H372-H341-H314-H318-H317
May intensify fire
oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.

P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c

H272-H314-H317-H334-H335-H341-H350-H360-H372

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

EINECSNumber : 237-124-1

RTECSNumber : SC9550000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents