missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) perchlorate hydrate, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 4431630

 View more versions of this product

Catalog No. AA4431630



Specifications

Specifications

Nickel (II) perchlorate hydrate
(metals basis)
Ni(ClO4)2·xH2O
UN1481
ZLQBNKOPBDZKDP-UHFFFAOYSA-L
nickel(2+);diperchlorate
26158
Crystalline
Odorless
Hygroscopic
140°C
13637-71-3
Cl2NiO8
MFCD00149804
nickel perchlorate, unii-bq8a24mezo, perchloric acid, nickel 2+ salt, nickel ii perchlorate, nickel 2+ diperchlorate, bq8a24mezo, perchloric acid, nickel 2+ salt 2:1, nickelous perchlorate, perchloric acid, nickel salt, nickel perchlorate ni clo4 2
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2]
257.585
257.59 (Anhydrous)
99.998%
1.55
Green
250g

Chemical Identifiers

13637-71-3
257.585
ZLQBNKOPBDZKDP-UHFFFAOYSA-L
26158
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2]
Cl2NiO8
MFCD00149804
nickel perchlorate, unii-bq8a24mezo, perchloric acid, nickel 2+ salt, nickel ii perchlorate, nickel 2+ diperchlorate, bq8a24mezo, perchloric acid, nickel 2+ salt 2:1, nickelous perchlorate, perchloric acid, nickel salt, nickel perchlorate ni clo4 2
nickel(2+);diperchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H334-H350-H360-H372-H341-H314-H318-H317
May intensify fire
oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

RTECSNumber : SC9550000

TSCA : TSCA

SDS Documents