missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) oxide, 99.99% (metals basis) , Puratronic™, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1081906

 View more versions of this product

Catalog No. AA1081906



Specifications

Specifications

Nickel(II) oxide
1313-99-1
NiO
14,6512
GNRSAWUEBMWBQH-UHFFFAOYSA-N
oxonickel
14805
Powder
6.67
Green
5g
Puratronic™
(metals basis)
MFCD00011145
black nickel oxide
O=[Ni]
74.692
74.71
99.998%
Odorless
1984°C

Chemical Identifiers

1313-99-1
74.692
GNRSAWUEBMWBQH-UHFFFAOYSA-N
14805
O=[Ni]
NiO
MFCD00011145
black nickel oxide
oxonickel
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317;
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : QR8400000

TSCA : TSCA

Documents