Please login to your online account to display your discounted pricing

Nickel(II) oxide, Puratronic™, 99.998% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Nickel(II) oxide
CAS 1313-99-1
Name Note Puratronic™
Molecular Formula NiO
Formula Weight 74.71
MDL Number MFCD00011145
Merck Index 14,6512
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1081906

  SDS
alfa aesar
1081906
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 74.71
Quantity 5g
Color Dark Green
Merck Index 14,6512
Health Hazard 2 GHS H Statement
H350-H372-H317;
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Melting Point 1984°C
Assay 99.998%
TSCA TSCA
MDL Number MFCD00011145
Physical Form Powder
RTECS Number QR8400000
Density 6.67
Odor Odorless
CAS 1313-99-1
Name Note Puratronic™
Molecular Formula NiO
Health Hazard 3 GHS P Statement
P260-P261-P280-P363-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay Percent Notes (metals basis)
Chemical Name or Material Nickel(II) oxide
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P363-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H372-H317;
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.