missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) oxide, Green, Ni 78.5%, Alfa Aesar™

Manufacturer:  Alfa Aesar 45094A1

 View more versions of this product

Catalog No. AA45094A1



Specifications

Specifications

Nickel(II) oxide
Ni 78.5%
MFCD00011145
GNRSAWUEBMWBQH-UHFFFAOYSA-N
oxonickel
14805
Powder
Green
1313-99-1
NiO
nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide, nickel ii oxide
O=[Ni]
74.692
74.71
Odorless
1kg

Chemical Identifiers

1313-99-1
74.692
GNRSAWUEBMWBQH-UHFFFAOYSA-N
14805
O=[Ni]
NiO
MFCD00011145
nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide, nickel ii oxide
oxonickel
Safety and Handling

Safety and Handling

H350-H372-H317-H413: May cause cancer. Causes damage to organs through prolonged or repeated exposure. May cause an allergic skin reaction. May cause long lasting harmful effects to aquatic life.

EINECSNumber : 215-215-7

SDS Documents