missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) oxide, black, Ni 76%, (contains Ni2O3)

Manufacturer:  Alfa Aesar™ 87302A1

 View more versions of this product

Catalog No. AA87302A1



Specifications

Specifications

Nickel (II) oxide
NiO
14,6512
O=[Ni]
74.692
74.71
Ni 76%, (contains Ni2O3)
Odorless
1984°C
1313-99-1
MFCD00011145
GNRSAWUEBMWBQH-UHFFFAOYSA-N
oxonickel
14805
Powder
6.67g/mL
Black
1kg

Chemical Identifiers

1313-99-1
74.692
GNRSAWUEBMWBQH-UHFFFAOYSA-N
oxonickel
NiO
MFCD00011145
14805
O=[Ni]
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : QR8400000

TSCA : TSCA

SDS Documents