missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) oxide, 99.995% (metals basis)

Vacuum deposition

Manufacturer:  Alfa Aesar™ 4442314

 View more versions of this product

Catalog No. AA4442314



Specifications

Specifications

Nickel (II) oxide
(metals basis)
MFCD00011145
black nickel oxide
GNRSAWUEBMWBQH-UHFFFAOYSA-N
oxonickel
14805
3-12mm Pieces
6.67 g/mL
3 to 12mm
1984°C
1313-99-1
NiO
14,6512
Soluble in ammonium hydroxide. Insoluble in water.
O=[Ni]
74.692
74.71
99.995%
Odorless
Black to Green
25g

Chemical Identifiers

1313-99-1
74.692
GNRSAWUEBMWBQH-UHFFFAOYSA-N
14805
O=[Ni]
NiO
MFCD00011145
black nickel oxide
oxonickel
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350i-H372

EINECSNumber : 215-215-7

RTECSNumber : QR8400000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents