missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) oxide, 99.99% (metals basis) , Puratronic™, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1081914

 View more versions of this product

Catalog No. AA1081914



Specifications

Specifications

Nickel(II) oxide
99.99% (metals basis)
MFCD00011145
GNRSAWUEBMWBQH-UHFFFAOYSA-N
oxonickel
14805
Powder
25g
1313-99-1
NiO
black nickel oxide
O=[Ni]
74.692
74.71
6.67

Chemical Identifiers

1313-99-1
74.692
GNRSAWUEBMWBQH-UHFFFAOYSA-N
14805
O=[Ni]
NiO
MFCD00011145
black nickel oxide
oxonickel
Documents