Learn More
Alfa Aesar™ Nickel(II) nitrate hexahydrate, tech., Ni 19.8% min
Nickel plating, manufacturing of ceramic colors
Manufacturer: Alfa Aesar™ 1222236
Specifications
Nickel(II) nitrate hexahydrate | |
H12N2NiO12 | |
MFCD00149805 | |
14,6513 | |
Soluble in water, alcohol | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ni+2] | |
290.791 | |
290.80 (182.70 Anhydrous) | |
Technical | |
2.05g/mL | |
Hygroscopic | |
Green | |
500g |
13478-00-7 | |
Ni(NO3)2·6H2O | |
UN2725 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
nickel(2+);dinitrate;hexahydrate | |
61630 | |
Flakes | |
Ni 19.8% min. | |
Odorless | |
137°C | |
56.7°C |
Chemical Identifiers
13478-00-7 | |
290.791 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
61630 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ni+2] |
H12N2NiO12 | |
MFCD00149805 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
nickel(2+);dinitrate;hexahydrate |
Safety and Handling
GHS H Statement
H272-H334-H350-H360-H372-H341-H318-H302-H332-H315-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
GHS P Statement
P221-P273-P201-P308+P313-P501a
Take any precaution to avoid mixing with combustibles.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NICKEL NITRATE
RTECSNumber : QR7300000
TSCA : TSCA