missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) iodide, anhydrous, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 2289306

 View more versions of this product

Catalog No. AA2289306



Specifications

Specifications

Nickel(II) iodide
13462-90-3
(metals basis)
MFCD00016264
BFSQJYRFLQUZKX-UHFFFAOYSA-L
diiodonickel
26038
5.834g/mL
Odorless
797°C
Anhydrous
99.5%
I2Ni
UN3077
[Ni](I)I
312.502
312.51
−8 Mesh
Light Sensitive & Hygroscopic
5g

Chemical Identifiers

13462-90-3
312.502
BFSQJYRFLQUZKX-UHFFFAOYSA-L
diiodonickel
I2Ni
MFCD00016264
26038
[Ni](I)I
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

SDS Documents