missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) hydroxide, Ni 61%

Manufacturer:  Alfa Aesar™ 12517A1

 View more versions of this product

Catalog No. AA12517A1



Specifications

Specifications

Nickel(II) hydroxide
Ni(OH)2
UN3077
Powder
4.1g/mL
Green
1kg
12054-48-7
MFCD00011140
92.7
Ni 61%
Odorless
230°C (decomposition)

Chemical Identifiers

12054-48-7
MFCD00011140
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P261-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : QR7040000

TSCA : TSCA

Documents