Please login to your online account to display your discounted pricing

Nickel(II) hydroxide, for analysis, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  275020050

 View more versions of this product

Catalog No. AC275020050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 92.7
Quantity 5g
Color Green
EINECS Number 235-008-5
TSCA TSCA
Purity Grade For analysis
Assay 61.0 to 65.0% (Nickel)
RTECS Number QR7040000
Merck Index 15, 6595
Iron (Fe) 200ppm max.
Manganese (Mn) 50ppm max.
Molecular Formula H2NiO2
Linear Formula Ni(OH)2
Sulfur (S) 50ppm max.
MDL Number MFCD00011140
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May damage the unborn child.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause allergy or asthma symptoms or breath
Copper (Cu) 10ppm max.
Chemical Name or Material Nickel(II) hydroxide
Name Note c.p., 60-61% Ni
CAS 12054-48-7
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Cobalt (Co) 8000ppm max.

Specifications

Chemical Name or Material Nickel(II) hydroxide
Name Note c.p., 60-61% Ni
CAS 12054-48-7
Purity Grade For analysis
Assay 61.0 to 65.0% (Nickel)
Molecular Formula H2NiO2
Linear Formula Ni(OH)2
Formula Weight 92.7
MDL Number MFCD00011140
Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage the unborn child.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause allergy or asthma symptoms or breath

SDS & Safety Documents