missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) fluoride tetrahydrate, 98+%

Manufacturer:  Alfa Aesar™ 1413218

 View more versions of this product

Catalog No. AA1413218



Specifications

Specifications

Nickel(II) fluoride tetrahydrate
MFCD00016263
168.77 (96.71 Anhydrous)
≥98%
50g
13940-83-5
UN3288
Powder
Odorless

Chemical Identifiers

13940-83-5
MFCD00016263
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : QR6825000

TSCA : TSCA

Documents