missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 1081522

 View more versions of this product

Catalog No. AA1081522



Specifications

Specifications

Nickel(II) chloride hydrate
(metals basis)
NiCl2·xH2O
UN3288
cl2ni.h2o
O.Cl[Ni]Cl
147.608
129.60 (Anhydrous)
99.995%
Hygroscopic
100g
69098-15-3
Cl2H2NiO
MFCD00149808
14,6505
HGGYAQHDNDUIIQ-UHFFFAOYSA-L
dichloronickel;hydrate
16212333
Crystalline Aggregates
Puratronic™
Green

Chemical Identifiers

69098-15-3
147.608
HGGYAQHDNDUIIQ-UHFFFAOYSA-L
16212333
O.Cl[Ni]Cl
Cl2H2NiO
MFCD00149808
cl2ni.h2o
dichloronickel;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H334-H350-H360-H372-H341-H315-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P273-P201-P309+P311
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : QR6480000

TSCA : TSCA

Documents