Learn More
Nickel(II) chloride hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04318514
Description
Used in the manufacture of all kinds of nickel compounds, preservatives and chemical reaction catalyst. Also used in electroplating, analytical chemistry, used for analysis reagent and in gas masks to absorb NH3. It has also been utilized in environmental studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel(II) chloride hexahydrate | |
Green | |
Crystalline | |
99.999% (Metals Basis) | |
MFCD00149809 | |
237.71 (129.60 Anhydrous) | |
1.92 |
140°C-H2O | |
7791-20-0 | |
25 g | |
Cl2Ni·6H2O | |
Soluble in water (2540g/L) and ethanol. | |
Odorless |
Chemical Identifiers
7791-20-0 | |
Cl2Ni·6H2O | |
MFCD00149809 |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c
H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 231-743-0
RTECSNumber : QR6480000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only