Learn More
Nickel(II) chloride hexahydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 207670250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel(II) chloride hexahydrate | |
7791-20-0,7718-54-9 | |
96.0 | |
4 to 6 (5% soln. at 20°C) | |
25 g | |
Cl2Ni·6H2O | |
MFCD00149809 | |
15, 6590 | |
237.71 | |
97% |
97% | |
Green | |
100.0 | |
Crystalline Powder and/or Crystals | |
96% min. (Complexometry) | |
NiCl2·6H2O | |
14,351; 15,96; 17,86 | |
Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol | |
237.71 | |
Glass bottle |
Chemical Identifiers
7791-20-0 | |
Cl2Ni·6H2O | |
237.71 | |
MFCD00149809 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid relea
GHS Signal Word: Danger
RTECSNumber : QR6480000
RUO – Research Use Only