Learn More
Nickel(II) chloride, anhydrous, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2208536
Description
Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.
Solubility
Soluble in ethylene glycol, ethanol, hydrazine and ammonium hydroxide. Slightly soluble in water. Insoluble in nitric acid.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with peroxides.
Specifications
Nickel(II) chloride | |
anhydrous | |
500 g | |
NiCl2 | |
UN3288 | |
Soluble in ethylene glycol,ethanol,hydrazine and ammonium hydroxide. Slightly soluble in water. Insoluble in nitric acid. | |
Cl[Ni]Cl | |
129.593 | |
CHEBI:34887 | |
98% | |
Hygroscopic |
1,001°C | |
7718-54-9 | |
Cl2Ni | |
MFCD00011142 | |
14,6505 | |
QMMRZOWCJAIUJA-UHFFFAOYSA-L | |
dichloronickel | |
24385 | |
129.6 | |
Odorless | |
3.55 |
Chemical Identifiers
7718-54-9 | |
129.593 | |
QMMRZOWCJAIUJA-UHFFFAOYSA-L | |
CHEBI:34887 | |
Cl[Ni]Cl |
Cl2Ni | |
MFCD00011142 | |
24385 | |
dichloronickel |
Safety and Handling
GHS H Statement
H301-H331-H334-H350-H360-H372-H341-H315-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c
H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 231-743-0
RTECSNumber : QR6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only