missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) chloride, 0.5M aq. soln.

$56.26 - $84.07

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2Ni
MDL Number MFCD00011142
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61204AD View Documents Alfa Aesar™
J61204AD
50mL Each for $56.26
 
AAJ61204AE View Documents Alfa Aesar™
J61204AE
100mL Each for $84.07
 
Specifications

Chemical Identifiers

7791-20-0
Cl2Ni
MFCD00011142

Specifications

7791-20-0
NiCl2
UN3082
Liquid
Odorless
50mL
Cl2Ni
MFCD00011142
129.6
0.5M aq. soln.
Green
Nickel(II) chloride
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H302-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H331-H334-H341-H350i-H360D-H372

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures