missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) chloride, 0.5M aq. soln.

Manufacturer:  Alfa Aesar™ J61204AE

 View more versions of this product

Catalog No. AAJ61204AE



Specifications

Specifications

7791-20-0
NiCl2
UN3082
Liquid
Odorless
100mL
Cl2Ni
MFCD00011142
129.6
0.5M aq. soln.
Green

Chemical Identifiers

7791-20-0
Cl2Ni
MFCD00011142
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H302-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H331-H334-H341-H350i-H360D-H372

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents