missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) carbonate hydroxide hydrate, for analysis, (ca 45-47% Ni), ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC223162500

 View more versions of this product

Catalog No. AC223162500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Nickel(II) carbonate hydroxide hydrate
39430-27-8,65405-96-1
Powder
CH4Ni3O7·xH2O
304.11
13, 6527
1000ppm max.
50ppm max.
100ppm max.
Plastic bottle
Green
ca 45-47% Ni, For analysis
For analysis
≥45% (NI)
2Ni(OH)2·xH2O
MFCD00150262
Basic nickel carbonate
1000ppm max.
0.05% max. (in HCl)
50ppm max.
50ppm max.
250g

Chemical Identifiers

39430-27-8,65405-96-1
2Ni(OH)2·xH2O
MFCD00150262
Nickel(II) carbonate hydroxide hydrate, ca 45-47% Ni, For analysis
For analysis
CH4Ni3O7·xH2O
304.11
Basic nickel carbonate
≥45% (NI)

Safety and Handling

SDS & Safety Documents

GHS H Statement
Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effects. May cause cancer by inhalation. Suspected of causing genetic defects. May damage the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
Wear eye protection/face protection. Use personal protective equipment as required. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

Danger