Please login to your online account to display your discounted pricing

Nickel(II) carbonate hydroxide hydrate, for analysis, (ca 45-47% Ni), ACROS Organics™


Quantity 1kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC223160010

 View more versions of this product

Catalog No. AC223160010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effects. May cause cancer by inhalation. Suspected of causing genetic defects. May damage the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Health Hazard 3 GHS P Statement
Wear eye protection/face protection. Use personal protective equipment as required. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Assay ≥45% (NI)
Formula Weight 304.11
Physical Form Powder
Quantity 1kg
Color Green
Merck Index 13, 6527
Purity Grade For analysis
Lead (Pb) 50ppm max.
Synonym Basic nickel carbonate
Trace Analysis 50ppm max.
Linear Formula 2Ni(OH)2·xH2O
Chemical Name or Material Nickel(II) carbonate hydroxide hydrate
Iron (Fe) 100ppm max.
Name Note ca 45-47% Ni, For analysis
CAS 39430-27-8
MDL Number MFCD00150262
CAS 65405-96-1
Copper (Cu) 50ppm max.
Chlorine (Cl) 1000ppm max.
Packaging Plastic bottle
Molecular Formula CH4Ni3O7·xH2O
Insoluble Matter 0.05% max. (in HCl)
Cobalt (Co) 1000ppm max.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Nickel(II) carbonate hydroxide hydrate
Name Note ca 45-47% Ni, For analysis
CAS 39430-27-8
CAS 65405-96-1
Purity Grade For analysis
Assay ≥45% (NI)
Molecular Formula CH4Ni3O7·xH2O
Linear Formula 2Ni(OH)2·xH2O
Formula Weight 304.11
MDL Number MFCD00150262
Synonym Basic nickel carbonate
Accessories

Danger

GHS P Statement
Wear eye protection/face protection. Use personal protective equipment as required. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS H Statement
Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effects. May cause cancer by inhalation. Suspected of causing genetic defects. May damage the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents