missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nickel(II) Carbonate, ACROS Organics™
Manufacturer: Fisher Scientific Acros 611485000
Specifications
Nickel(II) carbonate | |
CNiO3 | |
MFCD00011144 | |
ZULUUIKRFGGGTL-UHFFFAOYSA-L | |
nickel(2+);carbonate | |
18746 | |
Pass Test |
3333-67-3 | |
NiCO3 | |
carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate, nickel carbonate nico3, nickel ii carbonate, nickel monocarbonate, nickelous carbonate, unii-wm50squ829 | |
C(=O)([O-])[O-].[Ni+2] | |
118.701 | |
118.72 | |
500g |
Chemical Identifiers
3333-67-3 | |
118.701 | |
ZULUUIKRFGGGTL-UHFFFAOYSA-L | |
18746 | |
C(=O)([O-])[O-].[Ni+2] |
CNiO3 | |
MFCD00011144 | |
carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate, nickel carbonate nico3, nickel ii carbonate, nickel monocarbonate, nickelous carbonate, unii-wm50squ829 | |
nickel(2+);carbonate |
Safety and Handling
EINECSNumber : 222-068-2
RTECSNumber : QR6200000
TSCA : TSCA