missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nickel(II) Carbonate, Thermo Scientific Chemicals
Supplier: Fisher Scientific Acros 611481000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel(II) carbonate | |
100g | |
NiCO3 | |
carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate, nickel carbonate nico3, nickel ii carbonate, nickel monocarbonate, nickelous carbonate, unii-wm50squ829 | |
[Ni++].[O-]C([O-])=O | |
118.70 | |
118.72 |
3333-67-3 | |
CNiO3 | |
MFCD00011144 | |
ZULUUIKRFGGGTL-UHFFFAOYSA-L | |
nickel(2+) carbonate | |
18746 | |
Pass Test |
Chemical Identifiers
3333-67-3 | |
118.70 | |
ZULUUIKRFGGGTL-UHFFFAOYSA-L | |
18746 | |
[Ni++].[O-]C([O-])=O |
CNiO3 | |
MFCD00011144 | |
carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate, nickel carbonate nico3, nickel ii carbonate, nickel monocarbonate, nickelous carbonate, unii-wm50squ829 | |
nickel(2+) carbonate |
Safety and Handling
EINECSNumber : 222-068-2
RTECSNumber : QR6200000
TSCA : TSCA
RUO – Research Use Only