missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nickel(II) Carbonate, ACROS Organics™
$65.66 - $65.66
Chemical Identifiers
CAS | 3333-67-3 |
---|---|
Molecular Formula | CNiO3 |
Molecular Weight (g/mol) | 118.701 |
MDL Number | MFCD00011144 |
InChI Key | ZULUUIKRFGGGTL-UHFFFAOYSA-L |
Synonym | carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate, nickel carbonate nico3, nickel ii carbonate, nickel monocarbonate, nickelous carbonate, unii-wm50squ829 |
PubChem CID | 18746 |
IUPAC Name | nickel(2+);carbonate |
SMILES | C(=O)([O-])[O-].[Ni+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC611481000
|
Fisher Scientific Acros
611481000 |
100g |
Each for $65.66
|
|
AC611485000
|
Fisher Scientific Acros
611485000 |
500g |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
3333-67-3 | |
118.701 | |
ZULUUIKRFGGGTL-UHFFFAOYSA-L | |
18746 | |
C(=O)([O-])[O-].[Ni+2] |
CNiO3 | |
MFCD00011144 | |
carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate, nickel carbonate nico3, nickel ii carbonate, nickel monocarbonate, nickelous carbonate, unii-wm50squ829 | |
nickel(2+);carbonate |
Specifications
3333-67-3 | |
NiCO3 | |
carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate, nickel carbonate nico3, nickel ii carbonate, nickel monocarbonate, nickelous carbonate, unii-wm50squ829 | |
C(=O)([O-])[O-].[Ni+2] | |
118.701 | |
118.72 | |
100g |
CNiO3 | |
MFCD00011144 | |
ZULUUIKRFGGGTL-UHFFFAOYSA-L | |
nickel(2+);carbonate | |
18746 | |
Pass Test | |
Nickel(II) carbonate |
Safety and Handling
EINECSNumber : 222-068-2
RTECSNumber : QR6200000
TSCA : TSCA