missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1081314

 View more versions of this product

Catalog No. AA1081314



Specifications

Specifications

Nickel(II) acetate tetrahydrate
(metals basis)
Ni(CH3COO)2·4H2O
UN3077
14,6499
OINIXPNQKAZCRL-UHFFFAOYSA-L
nickel(2+);diacetate;tetrahydrate
62601
Crystalline Aggregates
Puratronic™
Light
25g
6018-89-9
C4H14NiO8
MFCD00066973
3730764
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Ni+2]
248.841
248.84 (176.79 Anhydrous)
99.999%
1.744g/mL
Green

Chemical Identifiers

6018-89-9
248.841
OINIXPNQKAZCRL-UHFFFAOYSA-L
62601
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Ni+2]
C4H14NiO8
MFCD00066973
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
nickel(2+);diacetate;tetrahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P261-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : QR6126000

TSCA : TSCA

Documents