missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) acetate hydrate, >99%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1222330

 View more versions of this product

Catalog No. AA1222330



Specifications

Specifications

Nickel(II) acetate hydrate
99+%
MFCD00012451
acetic acid, nickel +2 salt, tetrahydrate
OINIXPNQKAZCRL-UHFFFAOYSA-L
nickel(2+);diacetate;tetrahydrate
62601
Crystalline
White
250g
6018-89-9
C4H14NiO8
Green crystals
Soluble in water,alcohol
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Ni+2]
248.841
176.79 (Anhydrous)
Light
(decomposition)

Chemical Identifiers

6018-89-9
248.841
OINIXPNQKAZCRL-UHFFFAOYSA-L
62601
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Ni+2]
C4H14NiO8
MFCD00012451
acetic acid, nickel +2 salt, tetrahydrate
nickel(2+);diacetate;tetrahydrate
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302+H332-H317-H334-H335-H341-H350i-H360D-H372

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 206-761-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents