Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) 2,4-pentanedionate hydrate

$91.51 - $91.51

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1252930

 
alfa aesar™
1252930
250g Each for $91.51

AA12529A1

 
alfa aesar™
12529-A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 250g
Boiling Point 220°C (11mmHg)
Melting Point ≈230°C (dec.)
Quantity 250g
Chemical Name or Material Nickel(II) 2,4-pentanedionate hydrate
CAS 120156-44-7
Physical Form Powder
Formula Weight 256.91g/mol (Anhydrous)
MDL Number MFCD00000024
Beilstein 4157970
Merck Index 14,6500
Synonym Nickel(II) acetylacetonate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SA2100000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.