missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) 2,4-pentanedionate, 95%

Catalyst and U.V. stabilizer

Manufacturer:  Alfa Aesar™ 1881136

 View more versions of this product

Catalog No. AA1881136



Specifications

Specifications

Nickel(II) 2,4-pentanedionate
Hygroscopic
~230°C (decomposition)
3264-82-2
MFCD00000024
14,6500
Soluble in water, alcohol, toluene
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Ni+2]
256.911
256.91
95%
Odorless
Green
500g
C10H14NiO4
4157970
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
BMGNSKKZFQMGDH-SYWGCQIGSA-L
nickel(2+);(E)-4-oxopent-2-en-2-olate
53384569
Powder

Chemical Identifiers

3264-82-2
256.911
BMGNSKKZFQMGDH-SYWGCQIGSA-L
53384569
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Ni+2]
C10H14NiO4
MFCD00000024
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
nickel(2+);(E)-4-oxopent-2-en-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

Danger

RTECSNumber : SA2100000

TSCA : TSCA

SDS Documents