Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel(II) 1,1,1-trifluoro-2,4-pentanedionate dihydrate

$50.03 - $197.03

Chemical Identifiers

Chemical Name or Material Nickel(II) 1,1,1-trifluoro-2,4-pentanedionate dihydrate
CAS 55534-89-9
Formula Weight 400.90 (364.87 Anhydrous)
MDL Number MFCD00150656
Synonym Nickel(II) 1,1,1-trifluoroacetylacetonate dihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3944903

  SDS
alfa aesar™
3944903
1g Each for $50.03

AA3944906

  SDS
alfa aesar™
3944906
5g Each for $197.03
Description & Specifications

Specifications

Formula Weight 400.90 (364.87 Anhydrous)
Chemical Name or Material Nickel(II) 1,1,1-trifluoro-2,4-pentanedionate dihydrate
Physical Form Powder
MDL Number MFCD00150656
CAS 55534-89-9
Color Green
Quantity 1g
Synonym Nickel(II) 1,1,1-trifluoroacetylacetonate dihydrate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H302-H315-H319-H317
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Safety and Handling

Danger

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H350-H302-H315-H319-H317
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.