missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nickel ICP Standard, 1000 ppm Ni in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PNI1KN500
Specifications
Nickel ICP Standard | |
Natural Poly Bottle | |
93.0,4.76,0.09 | |
Green | |
Ni | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
1 mL = 1 mg Ni |
7440-02-0,7697-37-2,7440-02-0,7732-18-5 | |
500 mL | |
97.0,4.95,0.1 | |
1,000ppm Ni | |
MFCD00011137 MFCD06798735 | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Liquid |
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
CHEBI:28112 | |
[Ni] |
Ni | |
MFCD00011137 MFCD06798735 | |
935 | |
nickel |