Please login to your online account to display your discounted pricing

Nickel ICP Standard, 1mL = 1mg Ni (1,000ppm Ni), Ricca Chemical

$136.12 - $136.12

Chemical Identifiers

Chemical Name or Material Nickel ICP Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.76
CAS Max % 4.95
CAS 7440-02-0
CAS Min % 0.09
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PNI1KN-500

 
ricca chemical company
RIPNI1KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PNI1KN-100

 
ricca chemical company
RIPNI1KN100
Poly natural 100mL Each for $136.12
Description & Specifications

Specifications

Physical Form Liquid
Color Clear light green
Quantity 500mL
Chemical Name or Material Nickel ICP Standard
CAS 7732-18-5
Concentration or Composition Notes 1,000ppm Ni
Equivalence 1 mL = 1 mg Ni
CAS Min % 93.0
Packaging Poly natural
CAS Min % 4.76
CAS 7697-37-2
CAS Min % 0.09
CAS Max % 4.95
CAS Max % 0.1
CAS 7440-02-0
CAS Max % 97.0