missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nickel ICP Standard, 10,000 ppm Ni in 5% HNO3, Ricca Chemical
Chemical Identifiers
CAS | 7440-02-0 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
PNI10KN-50
|
Ricca Chemical Company
PNI10KN50 |
50 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PNI10KN-100
|
Ricca Chemical Company
PNI10KN100 |
100 mL | Natural LDPE Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PNI10KN-500
|
Ricca Chemical Company
PNI10KN500 |
500 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
CHEBI:28112 | |
[Ni] |
Ni | |
MFCD00011137 | |
935 | |
nickel |
Specifications
7440-02-0,7697-37-2,7440-02-0,7732-18-5 | |
50 mL | |
89.0,12.05,0.89 | |
10,000ppm Ni | |
MFCD00011137 | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Nickel ICP Standard |
Natural Poly Bottle | |
86.0,11.57,0.86 | |
Green | |
Ni | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
1 mL = 10 mg Ni |