Learn More
Nickel hydroxyacetate, Ni 27-29%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 3945614
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nickel hydroxyacetate | |
Green | |
Ni(OOCCH2OH)2 | |
25 g | |
nickel hydroxyacetate, acmc-20ak3m, nickel 2+ diglycolate, nickel 2+ bis hydroxyacetate, nickel 2+ ion diglycolate, bis hydroxyacetic acid nickel ii salt, nickel, 2,2'-bipyridine-n,n' hydroxyacetato 2--o1,o2-9ci | |
C(C(=O)[O-])O.C(C(=O)[O-])O.[Ni+2] | |
208.779 | |
208.78 | |
Powder |
41587-84-2 | |
C4H6NiO6 | |
MFCD00070414 | |
UN3077 | |
OZHLXQFAHIYYDJ-UHFFFAOYSA-L | |
2-hydroxyacetate;nickel(2+) | |
57349863 | |
Ni 27 to 29% |
Chemical Identifiers
41587-84-2 | |
208.779 | |
OZHLXQFAHIYYDJ-UHFFFAOYSA-L | |
57349863 | |
C(C(=O)[O-])O.C(C(=O)[O-])O.[Ni+2] |
C4H6NiO6 | |
MFCD00070414 | |
nickel hydroxyacetate, acmc-20ak3m, nickel 2+ diglycolate, nickel 2+ bis hydroxyacetate, nickel 2+ ion diglycolate, bis hydroxyacetic acid nickel ii salt, nickel, 2,2'-bipyridine-n,n' hydroxyacetato 2--o1,o2-9ci | |
2-hydroxyacetate;nickel(2+) |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302+H332-H317-H334-H341-H350-H360D-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures