missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel gauze, 100 mesh woven, Alfa Aesar™

Form: Wire Cloth; Open area: 36%; Width of opening: 0.006 in.

Manufacturer:  Thermo Fisher Scientific 44128GT

 View more versions of this product

Catalog No. AA44128GT



Specifications

Specifications

Nickel
7440-02-0
8.908g/cm3 at 20°C
MFCD00011137
carbonyl powder
[Ni]
Odorless
935
Wire Cloth
150 x 150mm
Silver gray guaze
Ni
100 Mesh
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
Silver to Gray

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
carbonyl powder
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H372

EINECSNumber : 231-111-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents