missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel foil, 2.0mm (0.08 in.) thick, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 44825HP

 View more versions of this product

Catalog No. AA44825HP



Specifications

Specifications

Nickel foil
100 x 500mm
7440-02-0
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
Odorless
nickel
935
Foil
≈44.5g/50x50mm
2.0mm (0.08 in.) thick
(metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
[Ni]
58.693
CHEBI:28112
99.5%

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

SDS Documents