missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Foil, 0.25mm (0.01 in.) thick, 99.5% (Metals basis), Alfa Aesar™

Form: Approx.5.6g/50 x 50mm

Manufacturer:  Alfa Aesar 44822GH

 View more versions of this product

Catalog No. AA44822GH



Specifications

Specifications

Nickel
7440-02-0
99.5% (Metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
[Ni]
58.693
CHEBI:28112
Silver to Gray
100 x 100mm
Silver gray foil
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
Odorless
nickel
935
Foil, 0.25mm (0.01 in.) thick

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

EINECSNumber : 231-111-4

SDS Documents