missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel foil, 0.025mm (0.001 in.) thick, annealed, 99% (metals basis), Alfa Aesar™

Form: approx. 13.6g/20x30cm, Temper: soft

Manufacturer:  Thermo Fisher Scientific 12722RX

 View more versions of this product

Catalog No. AA12722RX



Specifications

Specifications

Nickel
7440-02-0
99% (Metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
Silver to Gray
30 x 500cm
Silver gray foil
Ni
carbonyl powder
[Ni]
Odorless
935
Foil, 0.025mm (0.001 in.) thick

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
carbonyl powder
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H372

EINECSNumber : 231-111-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents