Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel dimethylglyoxime, 99%

Paint pigment

$109.28 - $109.28

Chemical Identifiers

Chemical Name or Material Nickel dimethylglyoxime
CAS 13478-93-8
Formula Weight 288.94
MDL Number MFCD00136028
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3945804

 
alfa aesar™
3945804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3945809

 
alfa aesar™
3945809
10g Each for $109.28

AA3945818

 
alfa aesar™
3945818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 288.94
Chemical Name or Material Nickel dimethylglyoxime
Physical Form Powder
Assay 99%
Melting Point 250°C (subl.)
MDL Number MFCD00136028
CAS 13478-93-8
Quantity 2g
Solubility Information Insoluble in water; Soluble in mineral acids
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H315-H319-H317-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H315-H319-H317-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.