Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Copper Iron Manganese rod, 1.27cm (0.5 in.) dia.

Chemical Identifiers

Chemical Name or Material Nickel Copper Iron Manganese rod
Name Note 1.27cm (0.5 in.) dia., cold drawn stress relieved, ASTM B164
MDL Number MFCD02091727
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36457BS

  SDS
alfa aesar™
36457BS
10cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36457BW

  SDS
alfa aesar™
36457BW
50cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Weight ≈104g/10cm
Name Note 1.27cm (0.5 in.) dia., cold drawn stress relieved, ASTM B164
Physical Form Rod
Health Hazard 2 GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Chemical Name or Material Nickel Copper Iron Manganese rod
MDL Number MFCD02091727
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10cm
Health Hazard 1 Warning
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cu:Fe:Mn; 63:28 to 34:2.5 max:2 max wt%
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.