Nickel Copper Foil, 0.51mm (0.02 in.) thick, Alfa Aesar™
Manufacturer:  Alfa Aesar 45148HB
 View more versions of this product
Catalog No. AA45148HB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Nickel Copper
Alloy 400
Ni:Cu; 67:33 wt.%
Foil, 0.5mm (0.02 in.) thick
300 x 300mm
Foil
MFCD00801098
Documents